Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human FGF-5 Protein

Click to view available options
Quantity:
1 mg
10 μg
50 μg
Description
An un-tagged bioactive recombinant protein corresponding to Human FGF-5
The Recombinant Human FGF-5 Protein has been validated for the following applications: SDS-Page, Bioactivity.
The Recombinant Human FGF-5 Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Specifications
Concentration | Lyoph |
For Use With (Application) | Bioactivity, SDS-PAGE |
Gene ID (Entrez) | 2250 |
Molecular Weight (g/mol) | 27.7 kDa |
Purification Method | >95%, by SDS-PAGE |
Quantity | 50 μg |
Source | E. coli |
Storage Requirements | Store at -20 to -80°C. Avoid freeze-thaw cycles. |
Gene Symbol | FGF5 |
Specific Reactivity | FGF-5 is a monomer, 27.7 kDa (252 aa) |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction