Learn More
Description
The Recombinant Human FGF-8 Animal-Free Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Specifications
| Concentration | Lyoph |
| For Use With (Application) | Bioactivity, SDS-PAGE |
| Gene ID (Entrez) | 2253 |
| Molecular Weight (g/mol) | 22.5 kDa |
| Purification Method | >95%, by SDS-PAGE |
| Quantity | 1 mg |
| Source | E. coli |
| Storage Requirements | Store at -20 to -80°C. Avoid freeze-thaw cycles. |
| Gene Symbol | FGF8 |
| Specific Reactivity | FGF-8 is a monomer, 22.5 kDa (194 aa) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
