Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human FGF-8 Protein
SDP

Catalog No. NB002067 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002067 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002067 Supplier Novus Biologicals™ Supplier No. NBP199966100UG

Highly purified and high bioactivity. Generating reliable and reproducible results.

Recombinant bioactive protein containing 193 amino acids for Human FGF-8 The Recombinant Human FGF-8 Protein is derived from E. coli. The Recombinant Human FGF-8 Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration Lyoph
For Use With (Application) Bioactivity, SDS-PAGE
Formulation Lyophilized from 10 mM Tris (pH 8.0), 75 mM NaCl.
Gene ID (Entrez) 2253
Molecular Weight (g/mol) 22.4 kDa
Quantity 100 μg
Source E. coli
Storage Requirements Store at -20 to -80C. Avoid freeze-thaw cycles.
Gene Alias AIGFKAL6, Androgen-induced growth factor, FGF-8, fibroblast growth factor 8, fibroblast growth factor 8 (androgen-induced), HBGF-8, Heparin-binding growth factor 8, MGC149376
Gene Symbol FGF8
Reconstitution Centrifuge the vial prior to opening. Reconstitute in 5 mM Tris (pH 8.0) to a concentration of 0.1 - 1 mg/mL. This solution can be diluted into other aqueous buffers and stored at 4°C for 1 week or -20°C for long-term storage.
Buffer Lyophilized from 10 mM Tris (pH 8.0), 75 mM NaCl.
Endotoxin Level <0.1 ng/μg
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.