Learn More
Description
The Recombinant Human FGF basic/FGF2 Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Specifications
| Concentration | Lyoph |
| For Use With (Application) | Bioactivity, SDS-PAGE |
| Gene ID (Entrez) | 2247 |
| Molecular Weight (g/mol) | 16.5 kDa |
| Purification Method | >95%, by SDS-PAGE |
| Quantity | 10 μg |
| Source | E. coli |
| Storage Requirements | Store at -20 to -80°C. Avoid freeze-thaw cycles. |
| Gene Symbol | FGF2 |
| Specific Reactivity | FGF basic/FGF2 is a monomer, 16.5 kDa (147 aa, Degredation of the full length FGF basic/FGF2 N-terminus results in a truncated 147aa protein. The N-terminus extensions influence the localization within the cell, but do not affect the biological activity) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
