Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Flt-3/Flk-2 His-tag Protein
Click to view available options
Quantity:
50 μg
Description
When Recombinant Human Flt-3/Flk-2 His-tag is captured onto a His Tag Antibody (Catalog # MAB050R ) coated plate, Recombinant Human Flt-3 Ligand (Catalog # 308-FK ) binds with an ED50 of 2-12 ng/mL.
Specifications
Specifications
| Accession Number | P36888 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 2322 |
| Molecular Weight (g/mol) | Observed: 82-93 kDa, reducing conditions; Theroretical: 59 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human Flt-3/Flk-2 protein Asn27-Asn541, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 500 μg/mL in PBS. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction