Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human FPRP/PTGFRN Fc Chimera Protein

Catalog No. 10433FP050
Click to view available options
Quantity:
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Measured by its binding ability in a functional ELISA.

When Recombinant Human FPRP/PTGFRN Fc Chimera (Catalog # 10433-FP) is immobilized at 5 μg/mL (100 μL/well), Biotinylated Recombinant Human CD9 Fc Chimera binds with an ED50 of 3-18 μg/mL.

Specifications

Accession Number Q9P2B2.2
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 5738
Molecular Weight (g/mol) Observed: 120-140 kDa, under reducing conditions; Theroretical: 117 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human FPRP/PTGFRN protein Human FPRP/PTGFRN (Arg22-Lys830) Accession # Q9P2B2.2(N-terminus) IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Protein FPRP/PTGFRN
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.