Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human G3BP1 His Protein
SDP

Catalog No. NBP15092550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15092550 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP15092550 Supplier Novus Biologicals™ Supplier No. NBP15092550UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-466 of Human G3BP1 The Recombinant Human G3BP1 Protein is derived from E. coli. The Recombinant Human G3BP1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number Q13283
For Use With (Application) In vitro Assay, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0) containing 10% glycerol, 2mM DTT, 100mM NaCl
Gene ID (Entrez) 10146
Molecular Weight (g/mol) 53.2kDa
Name G3BP1 Protein
Quantity 50 μg
Source E. Coli
Immunogen G3BP1, 1-466aa, Human, His tag, E.coli (NP_005745).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.