Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human gamma-Synuclein Protein
SDP

Catalog No. NBC118329 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118329 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118329 Supplier Novus Biologicals™ Supplier No. NBC118329
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-127 of Human gamma-Synuclein The Recombinant Human gamma-Synuclein Protein is derived from E. coli. The Recombinant Human gamma-Synuclein Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_003078
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 7.5), 0.1 M NaCl
Gene ID (Entrez) 6623
Molecular Weight (g/mol) 13.3kDa
Name Synuclein-gamma Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen MDVFKKGFSIAKEGVVGAVEKTKQGVTEAAEKTKEGVMYVGAKTKENVVQSVTSVAEKTKEQANAVSEAVVSSVNTVATKTVEEAENIAVTSGVVRKEDLRPSAPQQEGVASKEKEEVAEEAQSGGD
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1 microgram of protein (determined by LAL method)
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.