Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human GAP-43 Protein
SDP

Catalog No. NB253033100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB253033100 100 μg
NB25303320 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB253033100 Supplier Novus Biologicals™ Supplier No. NBP253033100UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-238 of Human GAP-43 The Recombinant Human GAP-43 Protein is derived from E. coli. The Recombinant Human GAP-43 Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 0.1M NaCl, 10% glycerol
Gene ID (Entrez) 2596
Molecular Weight (g/mol) 27.3kDa
Name Human GAP-43 Protein
Purification Method >90%, by SDS-PAGE
Quantity 100 μg
Source E.Coli
Immunogen MGSSHHHHHH SSGLVPRGSH MGSHMLCCMR RTKQVEKNDD DQKIEQDGIK PEDKAHKAAT KIQASFRGHI TRKKLKGEKK DDVQAAEAEA NKKDEAPVAD GVEKKGEGTT TAEAAPATGS KPDEPGKAGE TPSEEKKGEG DAATEQAAPQ APASSEEKAG SAETESATKA STDNSPSSKA EDAPAKEEPK QADVPAAVTA AAATTPAAED AAAKATAQPP TETGESSQAE ENIEAVDETK PKESARQDEG KEEEPEADQE HA
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias Axonal membrane protein GAP-43, B-50, calmodulin-binding protein P-57, growth associated protein 43, Growth-associated protein 43, nerve growth-related peptide GAP43, Neural phosphoprotein B-50, neuromodulin, neuron growth-associated protein 43, PP46, protein F1
Gene Symbol GAP43
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.