Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human GITR Ligand/TNFSF18 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Description
Measured by its ability to compete with biotinylated recombinant human GITR Ligand for binding to immobilized recombinant human GITR Fc Chimera.
Specifications
Specifications
| Accession Number | Q9UNG2 |
| For Use With (Application) | Binding Activity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 8995 |
| Molecular Weight (g/mol) | M.W. (Observed): 14 kDa-16 kDa, reducing conditions; M.W. (theoretical): 15 kDa |
| Name | GITR Ligand/TNFSF18 |
| Quantity | 25 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived human GITR Ligand/TNFSF18 protein Glu74-Ser199, with an N-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction