Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human GLP-1R Fc Chimera Protein
Click to view available options
:
50 μg
Description
Measured by its binding ability in a functional ELISA. When GLP-1 peptide (Tocris (Catalog # 5374 ) is immobilized at 0.25 μg/mL (100 μL/well), Recombinant Human GLP-1 Fc Chimera binds with an ED50 of 2-12 ng/mL.
Specifications
Specifications
Accession Number | P43220.2 |
For Use With (Application) | Bioactivity |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
Gene ID (Entrez) | 2740 |
Molecular Weight (g/mol) | M.W.-observed: 60-72 kDa, under reducing conditions, M.W.-theoretical: 40 kDa |
Quantity | 50 μg |
Source | Chinese Hamster Ovary cell line, CHO-derived human GLP-1 R protein Human GLP-1 R (Ala21&Arg24-Glu139;) (N-terminus) Accession # P43220.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
Gene Alias | GLP-1 receptor, GLP1R, GLP-1 R, GLP-1-R, glucagon-like peptide 1 receptor, MGC138331 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction