Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Glutathione Peroxidase 2/GPX2 His Protein
SDP

Catalog No. NBP17882410 Shop All R&D Systems Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17882410 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. NBP17882410 missing translation for 'mfr' Novus Biologicals™ missing translation for 'supplierNo' NBP17882410UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-190 of Human Glutathione Peroxidase 2/GPX2 The Recombinant Human Glutathione Peroxidase 2/GPX2 Protein is derived from E. coli. The Recombinant Human Glutathione Peroxidase 2/GPX2 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number P18283
For Use With (Application) SDS-PAGE
Formulation Liquid. 20mM Tris-HCl buffer (pH7.5) containing 40% glycerol, 0.15M NaCl, 1mM DTT
Gene ID (Entrez) 2877
Molecular Weight (g/mol) 24.1kDa
Name Glutathione Peroxidase 2/GPX2 Protein
Purification Method SDS-PAGE
Quantity 10 μg
Storage Requirements 4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.