Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human GP1BB His Protein

Click to view available options
Quantity:
10 μg
100 μg
20 μg
250 μg
50 μg
Specifications
Specifications
Concentration | 0.25 mg/mL |
For Use With (Application) | SDS-PAGE |
Formulation | PBS (pH 7.4), 10% glycerol |
Gene ID (Entrez) | 2812 |
Molecular Weight (g/mol) | 14 kDa |
Quantity | 20 μg |
Storage Requirements | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Endotoxin Concentration | < 1 EU per 1ug of protein (determined by LAL method) |
Gene Alias | antigen CD42b-beta, BDPLT1, BS, glycoprotein Ib (platelet), beta polypeptide, glycoprotein Ib platelet beta subunit, GPIBB, GPIbbeta, GPIb-beta, nuclear localization signal deleted in velocardiofacial syndrome, platelet glycoprotein Ib beta chain, platelet membrane glycoprotein Ib beta, truncated platelet membrane glycoprotein Ib beta |
Gene Symbol | GP1BB |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction