Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human GPT2 His Protein
SDP

Catalog No. NB171747 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
20 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171747 100 μg
NB171752 50 μg
NB171751 500 μg
NB171750 250 μg
NB171749 20 μg
NB171748 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. NB171747 Supplier Novus Biologicals™ Supplier No. NBP318182100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Specific activity is > 100units/mg, and is defined as the amount of enzyme that cleaves 1 umole of L-Alanine to L-Glutamate per minute at pH 7.5 at 37C

Specifications

Concentration 0.5 mg/ml
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 7.5), 30% glycerol, 2mM DTT, 0.2M NaCl
Gene ID (Entrez) 84706
Molecular Weight (g/mol) M.W. Theoretical: 60.3 kDa
Quantity 100 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Gene Alias AAT2, alanine aminotransferase 2, ALT2EC 2.6.1.2, Glutamate pyruvate transaminase 2, glutamic pyruvate transaminase (alanine aminotransferase) 2, Glutamic--alanine transaminase 2, glutamic-pyruvate transaminase 2, Glutamic--pyruvic transaminase 2, GPT 2
Gene Symbol GPT2
Research Category metabolism
Purity or Quality Grade >90%, by SDS-PAGE
Protein GPT2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.