Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Growth Hormone (GH) Protein

Catalog No. p-4719777
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Tris and NaCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Tris and NaCl.
This item is not returnable. View return policy

Catalog No. 1067GH025CF

Supplier: R&D Systems™ 1067GH025/CF

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Growth Hormone (GH) Protein is derived from E. coli. The Recombinant Human Growth Hormone (GH) Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 2688
Molecular Weight (g/mol) 22 kDa
Name Growth Hormone
Quantity 25 μg
Source E. coli-derived human Growth Hormone protein Phe27-Phe217,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias GH1, GHB5, GHIGHD1B, GHN, GH-N, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N, IGHD1A, IGHD2, Somatotropin
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.