Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human GTF2H5 His Protein
SDP

Catalog No. NBP25189105 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25189105 0.5 mg
NBP25189101 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25189105 Supplier Novus Biologicals™ Supplier No. NBP2518910.5MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-71 of Human GTF2H5 The Recombinant Human GTF2H5 Protein is derived from E. coli. The Recombinant Human GTF2H5 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation Phosphate buffered saline (pH7.4) containing 10% glycerol
Gene ID (Entrez) 404672
Molecular Weight (g/mol) TMW: 10.4kDa
Quantity 0.5 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias bA120J8.2, C6orf175trichothiodystrophy, FLJ30544, General transcription factor IIH polypeptide 5, general transcription factor IIH subunit 5, general transcription factor IIH, polypeptide 5, TFB5 ortholog, TFB5chromosome 6 open reading frame 175, TFIIH, TFIIH basal transcription factor complex TTDA subunit, TFIIH basal transcription factor complex TTD-A subunit, TTD, TTD-A, TTDATGF2H5
Common Name GTF2H5
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.