Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human HDAC3 Protein
SDP

Catalog No. NB002049 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002049 250 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002049 Supplier Novus Biologicals™ Supplier No. NBP199605250EACH

Highly purified and high bioactivity. Generating reliable and reproducible results.

Recombinant protein for Human HDAC3 The Recombinant Human HDAC3 Protein is derived from Sf 9 (baculovirus). The Recombinant Human HDAC3 Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration 0.5 mg/mL
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 25 mM Tris, pH 7.5, 100 mM NaCl, 2.7 mM KCl, 3 mM MgCl2, 10 % glycerol
Gene ID (Entrez) 8841
Molecular Weight (g/mol) 51.1 kDa
Quantity 250 Each
Source Sf 9 (baculovirus)
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Gene Alias EC 3.5.1.98, HD3RPD3-2RPD3, histone deacetylase 3, SMAP45
Gene Symbol HDAC3
Buffer 25 mM Tris, pH 7.5, 100 mM NaCl, 2.7 mM KCl, 3 mM MgCl2, 10 % glycerol.
Purity or Quality Grade >50% SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.