Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human His6- (DYKDDDDK)-p53 Protein
Click to view available options
:
20ug; Carrier Free
Description
The Recombinant Human His6-FLAG-p53 Protein is derived from E. coli.
Specifications
Specifications
Formulation | X mg/mL (X μM) in 50 mM HEPES pH 7.5, 450 mM NaCl, 10% Glycerol (v/v), 30 μM ZnCl 2 |
Gene ID (Entrez) | 7157 |
Name | His6-FLAG p53 |
Quantity | 20μg |
Source | E. coli-derived human p53 protein Contains an N-terminal 6-His and FLAG (DYKDDDDK) tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Purity or Quality Grade | >75%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction