Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Histamine N-Methyltransferase/HNMT Isoform 1 Protein
SDP

Catalog No. NBP2619805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
5 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2619805 5 μg
NBP26198020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP2619805 Supplier Novus Biologicals™ Supplier No. NBP2619805UG

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: SDS-Page, Bioactivity

A Biologically active recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-292 of Human Histamine N-Methyltransferase/HNMT The Recombinant Human Histamine N-Methyltransferase/HNMT Isoform 1 Protein is derived from E. coli. The Recombinant Human Histamine N-Methyltransferase/HNMT Isoform 1 Protein has been validated for the following applications: SDS-Page, Bioactivity.

Specifications

Concentration 1 mg/ml
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 20mM Tris-Hcl (pH8.0) buffer containing 10% glycerol
Gene ID (Entrez) 3176
Molecular Weight (g/mol) 37.4 kDa
Quantity 5 μg
Source E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol HNMT
Cross Reactivity Histamine N-Methyltransferase/HNMT
Species Human
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.