Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Histone H2B Protein
SDP

Catalog No. NB398082 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398082 200 μg
NB398081 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB398082 Supplier Novus Biologicals™ Supplier No. NBP307913200UG
Add to Cart
Add to Cart

Recombinant Protein

Specifications

For Use With (Application) Western Blot, SDS-PAGE, Kinase Assay
Formulation 1 x Dulbecco's PBS
Gene ID (Entrez) 440689
Molecular Weight (g/mol) MolecularWeight-theroretical: 15 kDa
Quantity 200 μg
Storage Requirements Store at -80°C. Avoid freeze/thaw cycles.
Gene Alias FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639
Gene Symbol HIST2H2BF
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Inhibitor Properties Can be used as a Western Blot control.
Form Purified
Research Category Chromatin Research, Epigenetics
Purity or Quality Grade >95%, by SDS-PAGE
Protein Histone H2B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.