Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Recombinant Human HLF His (C-Term) Protein
SDP

Catalog No. NB355234 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
20 μg
250 μg
50 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355234 20 μg
NB355233 250 μg
NB355232 50 μg
NB355236 100 μg
NB355235 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. NB355234 Supplier Novus Biologicals Supplier No. NBP32459820UG
Add to Cart
Add to Cart

Specifications

Concentration 0.5 mg/ml
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 3131
Molecular Weight (g/mol) 34 kDa
Quantity 20 μg
Storage Requirements Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration < 1 EU per 1ug of protein (determined by LAL method)
Gene Alias hepatic leukemia factor, MGC33822
Gene Symbol HLF
Purity or Quality Grade >95%, by SDS-PAGE
Protein HLF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.