Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human HPRT Protein
SDP

Catalog No. NB25306320 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB25306320 20 μg
NB253063100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB25306320 Supplier Novus Biologicals™ Supplier No. NBP25306320UG

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-218 of Human HPRT The Recombinant Human HPRT Protein is derived from E. coli. The Recombinant Human HPRT Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing, 20% glycerol
Gene ID (Entrez) 3251
Molecular Weight (g/mol) 26.7kDa
Name Human HPRT Protein
Purification Method >95%, by PAGE
Quantity 20 μg
Source E.Coli (Bioactivity Validated)
Immunogen MGSSHHHHHH SSGLVPRGSH MATRSPGVVI SDDEPGYDLD LFCIPNHYAE DLERVFIPHG LIMDRTERLA RDVMKEMGGH HIVALCVLKG GYKFFADLLD YIKALNRNSD RSIPMTVDFI RLKSYCNDQS TGDIKVIGGD DLSTLTGKNV LIVEDIIDTG KTMQTLLSLV RQYNPKMVKV ASLLVKRTPR SVGYKPDFVG FEIPDKFVVG YALDYNEYFR DLNHVCVISE TGKAKYKA
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias EC 2.4.2.8, HGPRTHPRTHGPRTase, hypoxanthine phosphoribosyltransferase 1, hypoxanthine-guanine phosphoribosyltransferase
Gene Symbol HPRT1
Biological Activity Specific activity is >15 units/mg and is defined as the amount of enzyme that catalyze the formation of 1μmole of guanosine 5-monophosphate(GMP) per minute from guanine and phosphoribosyl pyrophosphate at pH 7.5 at 37°C.
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.