Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human HSP60 His Protein
SDP

Catalog No. NB171986 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171986 100 μg
NB171987 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB171986 Supplier Novus Biologicals™ Supplier No. NBP318322100UG
This item is not returnable. View return policy

Recombinant Protein

The protein has ATPase activity at the time of manufacture of 3.6uM phosphate liberated/hr/ug protein in a 200ul reaction at 37C (pH7.5) in the presence of 20ul of 1mM ATP using a Malachite Green assay.

Specifications

For Use With (Application) Bioactivity, ELISA, Functional Assay, SDS-PAGE, Western Blot
Formulation 20mM Phosphate Buffer, 150mM NaCl, 10% glycerol
Gene ID (Entrez) 3329
Molecular Weight (g/mol) M.W. Theoretical: 60 kDa
Quantity 100 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias 60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant)
Gene Symbol HSPD1
Research Category Apoptosis, Cellular Markers, Loading Controls, Membrane Trafficking and Chaperones, Mitochondrial Markers
Purity or Quality Grade >85%
Protein HSP60
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.