Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human HSP90 beta Protein
SDP

Catalog No. NB171981 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171981 200 μg
NB171982 50 μg
NB171980 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. NB171981 Supplier Novus Biologicals™ Supplier No. NBP318320200UG
This item is not returnable. View return policy

Recombinant Protein

Specifications

For Use With (Application) Bioactivity, ELISA, Immunoprecipitation, SDS-PAGE, Surface Plasmon Resonance (SPR), Western Blot
Formulation 20mM Tris, pH7.5, 175 mM NaCl, 0.1 mM EDTA, 10% glycerol, 1 mM DTT
Gene ID (Entrez) 3326
Molecular Weight (g/mol) M.W. Theoretical: 90 kDa
Quantity 200 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Endotoxin Concentration Low Endotoxin
Gene Alias Heat shock 84 kDa, heat shock 90kD protein 1, beta, heat shock 90kDa protein 1, beta, heat shock protein 90kDa alpha (cytosolic), class B member 1, heat shock protein beta, heat shock protein HSP 90-beta, HSP 84, HSP84, HSP90-BETA, HSP90BHSP 90, HSPC2D6S182, HSPCBFLJ26984
Gene Symbol HSP90AB1
Research Category Cancer, Cellular Markers, ER Markers, Golgi Apparatus Markers, Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific
Purity or Quality Grade >85%
Protein HSP90 beta
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.