Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human HSP90 beta Protein
SDP

Catalog No. NB002042 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002042 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002042 Supplier Novus Biologicals™ Supplier No. NBP19957625UG

Highly purified. Generating reliable and reproducible results.

Recombinant bioactive protein for Human HSP90 beta The Recombinant Human HSP90 beta Protein is derived from Sf 9 (baculovirus). The Recombinant Human HSP90 beta Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 5 mg/mL
For Use With (Application) SDS-PAGE
Formulation 40 mM HEPES/KOH, pH 7.5, containing 400 mM potassium chloride and 5% glycerol
Gene ID (Entrez) 3326
Molecular Weight (g/mol) 83.1 kDa
Quantity 25 μg
Source Sf 9 (baculovirus)
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Gene Alias Heat shock 84 kDa, heat shock 90kD protein 1, beta, heat shock 90kDa protein 1, beta, heat shock protein 90kDa alpha (cytosolic), class B member 1, heat shock protein beta, heat shock protein HSP 90-beta, HSP 84, HSP84, HSP90-BETA, HSP90BHSP 90, HSPC2D6S182, HSPCBFLJ26984
Gene Symbol HSP90AB1
Buffer 40 mM HEPES/KOH, pH 7.5, containing 400 mM potassium chloride and 5% glycerol
Purity or Quality Grade >99%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.