Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human ICAM-5 hIgG-His Protein
SDP

Catalog No. NB398148 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398148 500 μg
NB398147 50 μg
NB398149 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB398148 Supplier Novus Biologicals™ Supplier No. NBP305774500UG
Add to Cart
Add to Cart

Recombinant Protein

Specifications

Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4) and 10% glycerol
Gene ID (Entrez) 7087
Molecular Weight (g/mol) MolecularWeight-theroretical: 113.8 kDa
Quantity 500 μg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Endotoxin Concentration Less Than 1 EU / 1 μg of protein (determined by LAL method)
Gene Alias ICAM-5, intercellular adhesion molecule 5, intercellular adhesion molecule 5, telencephalin, TLCNTelencephalin, TLNtelencephalin
Gene Symbol ICAM5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Form Purified
Research Category Immunology, Neuroscience
Purity or Quality Grade >90%, by SDS-PAGE
Protein ICAM-5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.