Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human ICOS (aa 21-134) His-tag Protein

Catalog No. 9865CS050
Click to view available options
Quantity:
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. Immobilized Recombinant Human B7-H2 Fc Chimera (Catalog # 165-B7) at 0.5 μg/mL (100 μL/well) can bind Recombinant Human ICOS with an ED50 of 25-125 ng/mL.

Specifications

Accession Number Q9Y6W8-1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 29851
Molecular Weight (g/mol) M.W. (Observed): 20-34 kDa, reducing conditions; M.W. (theoretical): 14 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human ICOS protein Human ICOS (Glu21-Gln134) (N-terminus) Accession # Q9Y6W8-1 HHHHHH IEGR (C-terminus)
Storage Requirements 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Cross Reactivity ICOS
Species Human
Reconstitution Reconstitute at 200 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein ICOS
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.