Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human IFIT3 His Protein

Click to view available options
Quantity:
0.1 mg
0.5 mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-490 of Human IFIT3 The Recombinant Human IFIT3 Protein is derived from E. coli. The Recombinant Human IFIT3 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Concentration | 1mg/mL |
For Use With (Application) | SDS-PAGE |
Formulation | Phosphate buffered saline (pH7.4) containing 20% glycerol, 1mM DTT |
Gene ID (Entrez) | 3437 |
Molecular Weight (g/mol) | TMW: 58.4kDa |
Quantity | 0.1 mg |
Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
Gene Alias | CIG-49, GARG-49, IFI60Retinoic acid-induced gene G protein, IFIT-3, IFIT-4, IFIT4IFI-60K, interferon-induced protein with tetratricopeptide repeats 3, Interferon-induced protein with tetratricopeptide repeats 4Interferon-induced 60 kDa protein, IRG2, ISG60, ISG-60, RIG-GCIG49 |
Common Name | IFIT3 |
Conjugate | Unconjugated |
Show More |
For research use only and is not approved for use in humans or in clinical diagnosis
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction