Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Human IFN-γ

Catalog No. p-4376449 Shop All BD Biosciences Products
Click to view available options
Quantity:
25 μg
50 μg
Concentration:
250μg/mL
500μg/mL

Pure by SDS-PAGE, and an absorbance assay based on the Beers-Lambert law

Frozen liquid comprised of 0.22μm sterile-filtered aqueous buffered solution and containing 2.5mg/ml biotechnology grade, low endotoxin bovine serum albumin, with no azide

  • Reactivity: Human
  • Storage Buffer: Frozen aqueous buffered solution containing BSA
  • Biological Activity: Activity range is measured in an anti-viral bioassay using A549 indicator cells
  • Specific Activity: 0.1 - 2.0 x 108 Units/mg
  • ED50: 0.05 to 1.0 ng/ml
  • Observed dose response relationship: 16X

Bioassay, enzyme-linked immunosorbent assay standard

TRUSTED_SUSTAINABILITY

Specifications

Concentration 250μg/mL
For Use With (Application) Bioassay, ELISA Standard
Formulation Frozen aqueous Buffered solution containing BSA
Molecular Weight (g/mol) 14 to 18
Name Human IFN-gamma Recombinant Protein
Purification Method SDS-PAGE
Quantity 25 μg
Storage Requirements -80°C
Cross Reactivity Human
Recombinant Recombinant
Form Solution
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.