Learn More
Description
Specifications
Specifications
| Accession Number | NM_024013 |
| Formulation | Sterile filtered and lyophilized with no additives. |
| Gene ID (Entrez) | 3439 |
| Name | IFN-alpha D Protein |
| Purification Method | SDS-PAGE |
| Quantity | 0.1 mg |
| Storage Requirements | −20°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Reconstitution | Reconstitute in H2O to a concentration of 0.1 - 1.0 mgml. The solution can then be diluted into other aqueous buffers and stored at 4C for 1 week or -20C for long-term storage. |
| Purity or Quality Grade | >95% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
