Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human IFN-alpha A Protein
Click to view available options
Quantity:
20 μg
5 μg
Description
The Recombinant Human IFN-alpha A Protein is derived from E. coli. The Recombinant Human IFN-alpha A Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Accession Number | V00549 |
For Use With (Application) | Bioactivity |
Formulation | Phosphate buffered saline (PBS) |
Molecular Weight (g/mol) | Predicted MW: 19.2kDa |
Name | IFN-alpha 2 |
Purification Method | A combination of ion exchange, hydrophobic interaction and size exclusion chromatography |
Quantity | 5 μg |
Source | E. coli-derived human IFN-alpha A protein |
Storage Requirements | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. |
Gene Alias | alpha-2a interferon, Ifa2, IFNA, IFNA2, IFNA2a, IFNA2b, IFNA2c, IFNAA, IFNalpha 2, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction