Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human IFN-alpha/beta R1 His Protein
SDP

Catalog No. NB002926
Click to view available options
Quantity:
0.05 mg
0.1 mg
0.5 mg

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a His-Tag and corresponding to the amino acids 28-436aa of Human IFN-alpha/beta R1.

Source: Baculovirus

Amino Acid Sequence:

KNLKSPQKVE VDIIDDNFIL RWNRSDESVG NVTFSFDYQK TGMDNWIKLS GCQNITSTKC NFSSLKLNVY EEIKLRIRAE KENTSSWYEV DSFTPFRKAQ IGPPEVHLEA EDKAIVIHIS PGTKDSVMWA LDGLSFTYSL VIWKNSSGVE ERIENIYSRH KIYKLSPETT YCLKVKAALL TSWKIGVYSP VHCIKTTVEN ELPPPENIEV SVQNQNYVLK WDYTYANMTF QVQWLHAFLK RNPGNHLYKW KQIPDCENVK TTQCVFPQNV FQKGIYLLRV QASDGNNTSF WSEEIKFDTE IQAFLLPPVF NIRSLSDSFH IYIGAPKQSG NTPVIQDYPL IYEIIFWENT SNAERKIIEK KTDVTVPNLK PLTVYCVKAR AHTMDEKLNK SSVFSDAVCE KTKPGNTSKH HHHHH>

Specifications

Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 3454
Molecular Weight (g/mol) 47.9 kDa
Quantity 0.1 mg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Alias alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFNBR, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1
Gene Symbol IFNAR1
Buffer PBS (pH 7.4), 10% glycerol.
Endotoxin Level < 1.0 EU per 1 μg of protein (determined by LAL method)
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.