Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-15 Protein

Catalog No. 247ilb500cf
Click to view available options
Quantity:
500 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using MO7e human megakaryocytic leukemic cells. The ED50 for this effect is 0.3-2.6 ng/mL. The specific activity of Recombinant Human IL-15 is approximately 4.5 x 105 U/μg, which is calibrated against recombinant human IL-15 WHO International Standard (NIBSC code: 95/554). Specific activity is for reference purposes only and is not routinely tested.

Specifications

Accession Number P40933.1
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 3600
Molecular Weight (g/mol) 10 kDa, under reducing conditions.
Quantity 500 μg
Source E. coli-derived human IL-15 protein Asn49-Ser162
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 °C to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Gene Alias IL15, IL-15MGC9721, interleukin 15, interleukin-15
Gene Symbol IL15
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 100μg/mL in sterile PBS.
Purity or Quality Grade >97%, by SDS-PAGE with silver staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.