Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human IL-17A (Human Cell-expressed) Protein
Click to view available options
Quantity:
10 μg
100 μg
Description
- Measured by its ability to induce CXCL1/GRO alpha secretion in HT-29 human colon adenocarcinoma cells.
- The ED50 for this effect is 0.12 to 1.2 ng/mL.
- Measured by its ability to induce IL-6 secretion by NIH-3T3 mouse embryonic fibroblast cells.
Specifications
Specifications
| Accession Number | Q16552 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 3605 |
| Molecular Weight (g/mol) | 15.1 kDa (monomer) |
| Name | CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A |
| Quantity | 100 μg |
| Source | Human embryonic kidney cell, HEK293-derived human IL-17/IL-17A protein Gly24-Ala155 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Gene Symbol | IL17A |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction