Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human IL-17F (NS0-expressed) Protein
Click to view available options
Quantity:
10 μg
100 μg
25 μg
Description
The Recombinant Human IL-17F (NS0-expressed) Protein is derived from NS0. The Recombinant Human IL-17F (NS0-expressed) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
Gene ID (Entrez) | 112744 |
Molecular Weight (g/mol) | 15.7 kDa (monomer) |
Name | IL-17F |
Quantity | 25 μg |
Source | Mouse myeloma cell line,NS0-derived human IL-17F protein Arg31-Gln163,with a C-terminal 6-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Structural Form | Disulfide-linked homodimer |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Show More |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction