Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human IL-1ra/IL-1F3 Protein
Click to view available options
Quantity:
10 μg
50 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Description
The Recombinant Human IL-1ra/IL-1F3 Protein is derived from E. coli. The Recombinant Human IL-1ra/IL-1F3 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
Gene ID (Entrez) | 3557 |
Molecular Weight (g/mol) | 17 kDa |
Name | IL-1ra/IL-1F3 |
Quantity | 50 μg |
Source | E. coli-derived human IL-1ra/IL-1F3 protein |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction