Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human IL-2 [ Cys Ser125]Protein
SDP

Catalog No. NB398135 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398135 100 μg
NB398132 500 μg
NB398134 10 μg
NB398133 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. NB398135 Supplier Novus Biologicals™ Supplier No. NBP307100100UG
Add to Cart
Add to Cart

Recombinant Protein

Fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine CTLL-2 cells is less than 0.1 ng/mL, corresponding to a specific activity of > 1.0 x 10^7 IU/mg.

Specifications

For Use With (Application) SDS-PAGE, Bioactivity
Formulation Lyophilized from a 0.2 mm filtered concentrated solution in 20 mM PB (pH 3.5).
Gene ID (Entrez) 3558
Molecular Weight (g/mol) MolecularWeight-theroretical: 15.5 kDa
Quantity 100 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 1.0 EU/μg of IL-2 AS determined by LAL method.
Gene Alias aldesleukin, IL-2, IL-2 lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF
Gene Symbol IL2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to 0.1-1.0 mg/mL Stock solutions should be apportioned into working aliquots and stored (at -20°C.)
Form Purified
Research Category Adaptive Immunity, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers
Purity or Quality Grade >97 % pure by SDS-PAGE and HPLC
Protein IL-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.