Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-7 (HEK-293-expressed) Protein

Catalog No. 11089IL01M
Click to view available options
Quantity:
1 mg
10 μg
250 μg
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Recombinant Protein

Measured in a cell proliferation assay using PHA-activated human peripheral blood lymphocytes (PBL). Yokota, T. et al. (1986) Proc. Natl. Acad. Sci. USA 83:5894. The ED50 for this effect is 0.100-0.500 ng/mL. The specific activity of Recombinant Human IL-7 is >1.00 x 108 units/mg, which is calibrated against the human IL-7 reference standard (NIBSC code: 90/530). Specific activity is for reference purposes only and is not routinely tested.

Specifications

Accession Number P13232.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3574
Molecular Weight (g/mol) MolecularWeight-observed: 18-28 kDa, under reducing conditions.; MolecularWeight-theroretical: 17 kDa
Quantity 1 mg
Source Human embryonic kidney cell, HEK293-derived human IL-7 protein Asp26 - His177
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias IL7, IL-7 interleukin-7, interleukin 7, Lymphopoietin-1, PBGF
Conjugate Unconjugated
Reconstitution Reconstitute at 100 - 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-7
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.