Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Indoleamine 2,3-dioxygenase/IDO Protein
SDP

Catalog No. NB253085100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB253085100 100 μg
NB25308520 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB253085100 Supplier Novus Biologicals™ Supplier No. NBP253085100UG

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-403 of Human Indoleamine 2,3-dioxygenase/IDO The Recombinant Human Indoleamine 2,3-dioxygenase/IDO Protein is derived from E. coli. The Recombinant Human Indoleamine 2,3-dioxygenase/IDO Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 0.15M NaCl, 20% glycerol, 1mM DTT
Gene ID (Entrez) 3620
Molecular Weight (g/mol) 47.7kDa
Name Human Indoleamine 2,3-dioxygenase/IDO Protein
Purification Method >90%, by SDS-PAGE
Quantity 100 μg
Source E.Coli (Bioactivity Validated)
Immunogen MGSSHHHHHH SSGLVPRGSH MGSMAHAMEN SWTISKEYHI DEEVGFALPN PQENLPDFYN DWMFIAKHLP DLIESGQLRE RVEKLNMLSI DHLTDHKSQR LARLVLGCIT MAYVWGKGHG DVRKVLPRNI AVPYCQLSKK LELPPILVYA DCVLANWKKK DPNKPLTYEN MDVLFSFRDG DCSKGFFLVS LLVEIAAASA IKVIPTVFKA MQMQERDTLL KALLEIASCL EKALQVFHQI HDHVNPKAFF SVLRIYLSGW KGNPQLSDGL VYEGFWEDPK EFAGGSAGQS SVFQCFDVLL GIQQTAGGGH AAQFLQDMRR YMPPAHRNFL CSLESNPSVR EFVLSKGDAG LREAYDACVK ALVSLRSYHL QIVTKYILIP ASQQPKENKT SEDPSKLEAK GTGGTDLMNF LKTVRSTTEK SLLKEG
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias EC 1.13.11.52, IDOIDO-1, INDOindole 2,3-dioxygenase, indoleamine 2,3-dioxygenase 1, indoleamine-pyrrole 2,3 dioxygenase, Indoleamine-pyrrole 2,3-dioxygenase
Gene Symbol IDO1
Biological Activity Specific activity is >300 pmol/min/μg, and is defined as the amount of enzyme that hydrolyze 1.0 pmole of L-Tryptophan to N-formyl-L-kynurenine per minute at pH 6.5 at 25°C.
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.