Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Integrin alpha V beta 1 Protein
Click to view available options
Quantity:
1 mg
50 μg
Description
The Recombinant Human Integrin alpha V beta 1 Protein is derived from CHO. The Recombinant Human Integrin alpha V beta 1 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
For Use With (Application) | Bioactivity |
Formulation | Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. |
Gene ID (Entrez) | 3685 |
Molecular Weight (g/mol) | 115 kDa ( alpha V subunit) and 86 kDa ( beta 1 subunit) |
Quantity | 1 mg |
Source | Chinese Hamster Ovary cell line,CHO-derived human Integrin alpha V beta 1 protein, (N-terminus) Human Integrin alpha V (Phe31-Val992) Accession &Num; NP_002201 GGGSGGGSGGGS Acidic Tail HHHHHH Human Integrin beta 1 (Gln21-Asp728) Accession &Num; P05556 GGGSGGGSGGGS Basic Tail (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Endotoxin Concentration | <1.0 EU per 1μg of the protein by the LAL method. |
Conjugate | Unconjugated |
Species | Human |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction