Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Integrin beta 4/CD104 Protein
SDP

Catalog No. NBP26146110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP26146110 10 μg
NBP26146150 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP26146110 Supplier Novus Biologicals™ Supplier No. NBP26146110UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a His-tag and corresponding to the amino acids 28-710 of Human Integrin beta 4/CD104. The Recombinant Human Integrin beta 4/CD104 Protein is derived from Baculovirus. The Recombinant Human Integrin beta 4/CD104 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5 mg/ml
For Use With (Application) SDS-PAGE
Formulation Phosphate Buffered Saline (pH 7.4) containing 10% glycerol.
Gene ID (Entrez) 3691
Molecular Weight (g/mol) 77.5 kDa
Quantity 10 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol ITGB4
Cross Reactivity Integrin beta 4/CD104
Species Human
Purity or Quality Grade >90%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.