Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human IRF3 Protein
SDP

Catalog No. NBC118469 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118469 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118469 Supplier Novus Biologicals™ Supplier No. NBC118469
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-112 of Human IRF3 The Recombinant Human IRF3 Protein is derived from E. coli. The Recombinant Human IRF3 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001562
For Use With (Application) ELISA, SDS-PAGE
Formulation PBS (pH 7.4)
Gene ID (Entrez) 3661
Molecular Weight (g/mol) 13kDa
Name IRF3 Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MGTPKPRILPWLVSQLDLGQLEGVAWVNKSRTRFRIPWKHGLRQDAQQEDFGIFQAWAEATGAYVPGRDKPDLPTWKRNFRSALNRKEGLRLAEDRSKDPHDPHKIYEFVNS
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.