Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human ISM1 Protein
Click to view available options
Quantity:
50 μg
Description
The Recombinant Human ISM1 Protein is derived from CHO. The Recombinant Human ISM1 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | B1AKI9 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 140862 |
| Molecular Weight (g/mol) | 50 kDa |
| Name | bA149I18.1, C20orf82, Chromosome 20 Open Reading Frame 82, dJ1077I2.1, ISM, ISM1, ISM-1, Isthmin, Isthmin 1 Homolog, Isthmin 1 Homolog (Zebrafish), isthmin-1 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Isthmin 1/ISM1 protein Ser27-Tyr464,with a C-terminal 6-His tag |
| Storage Requirements | 12 months from date of receipt, ≤ -20° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, ≤ -20° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction