Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human KIR3DS1/CD158e2 Fc Chimera Protein
Click to view available options
Quantity:
50 μg
Description
The Recombinant Human KIR3DS1/CD158e2 Fc Chimera Protein is derived from CHO. The Recombinant Human KIR3DS1/CD158e2 Fc Chimera Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
Gene ID (Entrez) | 3813 |
Molecular Weight (g/mol) | 61.9 kDa (monomer) |
Name | KIR3DS1/CD158e2 Fc Chimera |
Quantity | 50 μg |
Source | Chinese Hamster Ovary cell line,CHO-derived human KIR3DS1/CD158e2 protein, (N-terminus) Human KIR3DS1 (His22-His340) Accession &Num;Q14943 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Structural Form | Disulfide-linked homodimer |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction