Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Lefty-A Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Description
The Recombinant Human Lefty-A Protein is derived from NS0. The Recombinant Human Lefty-A Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 7044 |
| Molecular Weight (g/mol) | 39 kDa |
| Name | Lefty-A |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived human Lefty-A protein, (N-terminus) Mouse Lefty-1 Propeptide (Leu22-Arg77) Accession &Num; Q64280 HHHHHH Human Lefty-A (Phe78-Pro366) Accession &Num; O00292 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Reconstitution | Reconstitute at 500μg/mL in sterile PBS. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction