Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human LKB1/STK11 His Protein
SDP

Catalog No. NBP25195905 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25195905 0.05 mg
NBP25195925 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25195905 Supplier Novus Biologicals™ Supplier No. NBP2519590.05MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-433 of Human LKB1/STK11 The Recombinant Human LKB1/STK11 Protein is derived from E. coli. The Recombinant Human LKB1/STK11 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 10% Glycerol
Gene ID (Entrez) 6794
Molecular Weight (g/mol) TMW: 51kDa
Quantity 0.05 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias EC 2.7.11.1, LKB1 serine/threonine kinase 11 (Peutz-Jeghers syndrome), PJS, PJS polarization-related protein LKB1, Renal carcinoma antigen NY-REN-19, serine/threonine kinase 11, serine/threonine-protein kinase 11, Serine/threonine-protein kinase LKB1, STK11
Common Name LKB1/STK11
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.