Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human M-CSF Protein
SDP

Catalog No. nbp25958610
Click to view available options
Quantity:
10 μg
50 μg

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 33-255 of Human M-CSF The Recombinant Human M-CSF Protein is derived from Baculovirus. The Recombinant Human M-CSF Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Concentration 0.25mg/mL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid. PBS (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 1435
Molecular Weight (g/mol) 26.1 kDa
Name Recombinant Human M-CSF Protein
Purification Method >90%, by SDS-PAGE
Quantity 10 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of protein (determined by LAL method)
Gene Alias colony stimulating factor 1 (macrophage), CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, M-CSF, MCSFlanimostim, MGC31930
Gene Symbol CSF1
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.