Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human M-CSF R/CD115 Fc Avi-tag Protein
Click to view available options
Quantity:
50 μg
Description
The biotin to protein ratio is greater than 0.7 as determined by the HABA assay. Measured by its binding ability in a functional ELISA. When Human M-CSF (Catalog # 216-MC ) is immobilized at 1 μg/mL (100 μL/well), it binds to Biotinylated Recombinant Human M-CSF R/CD115 Fc Chimera Avi-tag (Catalog # AVI10339) with an ED50 of 6-36 ng/mL.
Specifications
Specifications
Accession Number | P07333-1 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
Gene ID (Entrez) | 1436 |
Molecular Weight (g/mol) | Observed: 100-120 kDa, under reducing conditions; Theroretical: 83 kDa |
Quantity | 50 μg |
Source | Chinese Hamster Ovary cell line, CHO-derived human M-CSF R/CD115 protein Human M-CSF R/CD115 (Ile20-Thr516) Accession # P07333.2(N-terminus) IEGRMD Human IgG1 (Pro100-Lys330) Avi-tag (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
Conjugate | Biotin |
Reconstitution | Reconstitute at 500 μg/mL in PBS. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction