Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human MECR His Protein
SDP

Catalog No. NBP259511Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259511Y 250 μg
NBP25951150 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP259511Y Supplier Novus Biologicals™ Supplier No. NBP259511250UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 54-373 of Human MECR The Recombinant Human MECR Protein is derived from E. coli. The Recombinant Human MECR Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. 20mM Tris-HCl buffer (pH8.0) containing 0.2M NaCl, 5mM DTT, 20% glycerol
Gene ID (Entrez) 51102
Molecular Weight (g/mol) 36.9 kDa
Name Recombinant Human MECR Protein
Purification Method >90%, by SDS-PAGE
Quantity 250 μg
Source E.coli
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias CGI-63, EC 1.3.1.38, FASN2B, homolog of yeast 2-enoyl thioester reductase, HsNrbf-1, mitochondrial 2-enoyl thioester reductase, mitochondrial trans-2-enoyl-CoA reductase, NBRF1, NRBF-1, NRBF1hsNrbf-1, nuclear receptor binding factor 1, Nuclear receptor-bi
Gene Symbol MECR
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.