Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human MMP-9 Protein
SDP

Catalog No. NB002048 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002048 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002048 Supplier Novus Biologicals™ Supplier No. NBP19960450UG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Highly purified. Generating reliable and reproducible results.

Recombinant protein for Human MMP-9 corresponding to the catalytic domain The Recombinant Human MMP-9 Protein is derived from E. coli. The Recombinant Human MMP-9 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration Lyoph
For Use With (Application) SDS-PAGE
Formulation Lyophilized with no additives.
Gene ID (Entrez) 4318
Molecular Weight (g/mol) 39.0 kDa
Quantity 50 μg
Source E. coli
Storage Requirements Store at -20 to -80C. Avoid freeze-thaw cycles.
Gene Alias 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase
Gene Symbol MMP9
Reconstitution Reconstitute in sterile PBS to 0.1 - 1.0 mg/mL. This solution can then be diluted into other aqueous buffers andstored at 4°C for 1 week or -20°C for long-term storage.
Buffer Lyophilized with no additives.
Endotoxin Level <0.1 ng/μg
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.